Anti-CASP7 Mouse monoclonal antibody

Lieferant: OriGene
ORIGTA301617EA 498 EUR
ORIGTA301617
Anti-CASP7 Mouse monoclonal antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR