Anti-SLC9A8 Mouse Monoclonal Antibody [clone: 7A11]

Lieferant: OriGene
ORIGTA301650EA 498 EUR
ORIGTA301650
Anti-SLC9A8 Mouse Monoclonal Antibody [clone: 7A11]
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: SLC9A8 (solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR