Anti-KCNK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301657EA 658 EUR
ORIGTA301657
Anti-KCNK2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: KCNK2 (potassium channel, subfamily K, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR