Anti-YWHAG Mouse Monoclonal Antibody [clone: HS23]

Lieferant: OriGene
ORIGTA301661EA 570 EUR
ORIGTA301661
Anti-YWHAG Mouse Monoclonal Antibody [clone: HS23]
Antikörper
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: YWHAG (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR