Anti-PDE4D Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA302565EA 547 EUR
ORIGTA302565
Anti-PDE4D Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: PDE4D (phosphodiesterase 4D, cAMP-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR