Anti-VANGL2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303193EA 536 EUR
ORIGTA303193
Anti-VANGL2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: VANGL2 (VANGL planar cell polarity protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR