Anti-ABCD1 Goat Antibody

Lieferant: OriGene
ORIGTA305801EA 547 EUR
ORIGTA305801
Anti-ABCD1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: ABCD1 (ATP-binding cassette, sub-family D (ALD), member 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR