Anti-TNKS2 Goat Antibody

Lieferant: OriGene
ORIGTA305802EA 547 EUR
ORIGTA305802
Anti-TNKS2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: TNKS2 (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR