Anti-SH2D1B Goat Antibody

Lieferant: OriGene
ORIGTA305804EA 550 EUR
ORIGTA305804
Anti-SH2D1B Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: SH2D1B (SH2 domain containing 1B)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR