Anti-MLPH Goat Antibody

Lieferant: OriGene
ORIGTA305809EA 547 EUR
ORIGTA305809
Anti-MLPH Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-2?g/ml Immunofluorescence: 05?g/ml

Type: Primary
Antigen: MLPH (melanophilin)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR