Anti-AGAP3 Goat Antibody

Lieferant: OriGene
ORIGTA305820EA 547 EUR
ORIGTA305820
Anti-AGAP3 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: AGAP3 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR