Anti-TH Goat Antibody

Lieferant: OriGene
ORIGTA305827EA 547 EUR
ORIGTA305827
Anti-TH Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4?g/ml

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR