Anti-LRP5 Goat Antibody

Lieferant: OriGene
ORIGTA305828EA 547 EUR
ORIGTA305828
Anti-LRP5 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 3-6?g/ml

Type: Primary
Antigen: LRP5 (low density lipoprotein receptor-related protein 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR