Anti-NTN1 Goat Antibody

Lieferant: OriGene
ORIGTA305829EA 550 EUR
ORIGTA305829
Anti-NTN1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: NTN1 (netrin 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR