Anti-CHAT Goat Antibody

Lieferant: OriGene
ORIGTA305830EA 550 EUR
ORIGTA305830
Anti-CHAT Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 02-05?g/ml

Type: Primary
Antigen: CHAT (choline O-acetyltransferase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR