Anti-DIO2 Goat Antibody

Lieferant: OriGene
ORIGTA305836EA 547 EUR
ORIGTA305836
Anti-DIO2 Goat Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 2-3?g/ml

Type: Primary
Antigen: DIO2 (deiodinase, iodothyronine, type II)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR