Anti-NME9 Goat Antibody

Lieferant: OriGene
ORIGTA305854EA 547 EUR
ORIGTA305854
Anti-NME9 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: NME9 (NME/NM23 family member 9)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR