Anti-SERPINA12 Goat Antibody

Lieferant: OriGene
ORIGTA305856EA 550 EUR
ORIGTA305856
Anti-SERPINA12 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: SERPINA12 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR