Anti-ABCA9 Goat Antibody

Lieferant: OriGene
ORIGTA305862EA 550 EUR
ORIGTA305862
Anti-ABCA9 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: ABCA9 (ATP-binding cassette, sub-family A (ABC1), member 9)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR