Anti-CACNA1C Goat Antibody

Lieferant: OriGene
ORIGTA305878EA 547 EUR
ORIGTA305878
Anti-CACNA1C Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: CACNA1C (calcium channel, voltage-dependent, L type, alpha 1C subunit)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR