Anti-ANKK1 Goat Antibody

Lieferant: OriGene
ORIGTA305881EA 550 EUR
ORIGTA305881
Anti-ANKK1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: ANKK1 (ankyrin repeat and kinase domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR