Drucken

Anti-LTF Goat Antibody

Lieferant: OriGene
ORIGTA305892EA 438 EUR InStock
ORIGTA305892
Anti-LTF Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 005-02?g/ml

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more