Anti-ITPKC Goat Antibody

Lieferant: OriGene
ORIGTA305898EA 547 EUR
ORIGTA305898
Anti-ITPKC Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ITPKC (inositol-trisphosphate 3-kinase C)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR