Anti-UNC5C Goat Antibody

Lieferant: OriGene
ORIGTA305907EA 550 EUR
ORIGTA305907
Anti-UNC5C Goat Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 50?g/ml

Type: Primary
Antigen: UNC5C (unc-5 homolog C (C. elegans))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR