Anti-PSMB10 Goat Antibody

Lieferant: OriGene
ORIGTA305923EA 547 EUR
ORIGTA305923
Anti-PSMB10 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: PSMB10 (proteasome (prosome, macropain) subunit, beta type, 10)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR