Anti-LIG1 Goat Antibody

Lieferant: OriGene
ORIGTA305925EA 547 EUR
ORIGTA305925
Anti-LIG1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: LIG1 (ligase I, DNA, ATP-dependent)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR