Anti-TNFRSF25 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305945EA 600 EUR
ORIGTA305945
Anti-TNFRSF25 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TNFRSF25 (tumor necrosis factor receptor superfamily, member 25)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR