Anti-MAP3K14 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305950EA 600 EUR
ORIGTA305950
Anti-MAP3K14 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR