Drucken

Anti-CFLAR Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305978EA 482 EUR InStock
ORIGTA305978
Anti-CFLAR Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000 Immunochemistry: 2 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CFLAR (CASP8 and FADD-like apoptosis regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more