Anti-RIPK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305984EA 600 EUR
ORIGTA305984
Anti-RIPK2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: RIPK2 (receptor-interacting serine-threonine kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR