Anti-CHUK Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305994EA 600 EUR
ORIGTA305994
Anti-CHUK Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: CHUK (conserved helix-loop-helix ubiquitous kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR