Drucken

Anti-IL1RAP Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306001EA 482 EUR InStock
ORIGTA306001
Anti-IL1RAP Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: IL1RAP (interleukin 1 receptor accessory protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more