Anti-STK17A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306006EA 595 EUR
ORIGTA306006
Anti-STK17A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: STK17A (serine/threonine kinase 17a)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR