Anti-BCL10 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306011EA 600 EUR
ORIGTA306011
Anti-BCL10 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5 μg/ml; Immunochemistry: 1 μg/ml; Immunofluorescence: 10 μg/ml

Type: Primary
Antigen: BCL10 (B-cell CLL/lymphoma 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR