Anti-TNFRSF10C Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306012EA 600 EUR
ORIGTA306012
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR