Anti-BNIP3L Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306045EA 600 EUR
ORIGTA306045
Anti-BNIP3L Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: BNIP3L (BCL2/adenovirus E1B 19kDa interacting protein 3-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR