Anti-IKBKG Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306055EA 600 EUR
ORIGTA306055
Anti-IKBKG Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR