Drucken

Anti-CD209 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306058EA 482 EUR InStock
ORIGTA306058
Anti-CD209 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CD209 (CD209 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more