Anti-IRAK3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306061EA 600 EUR
ORIGTA306061
Anti-IRAK3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: IRAK3 (interleukin-1 receptor-associated kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR