Anti-TNFRSF13B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306064EA 600 EUR
ORIGTA306064
Anti-TNFRSF13B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: TNFRSF13B (tumor necrosis factor receptor superfamily, member 13B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR