Anti-TP53AIP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306074EA 595 EUR
ORIGTA306074
Anti-TP53AIP1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 4-8 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TP53AIP1 (tumor protein p53 regulated apoptosis inducing protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR