Anti-IL22RA1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306082EA 600 EUR
ORIGTA306082
Anti-IL22RA1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IL22RA1 (interleukin 22 receptor, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR