Anti-BIRC7 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306089EA 600 EUR
ORIGTA306089
Anti-BIRC7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: BIRC7 (baculoviral IAP repeat containing 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR