Anti-PAK6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306114EA 595 EUR
ORIGTA306114
Anti-PAK6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-4 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: PAK6 (p21 protein (Cdc42/Rac)-activated kinase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR