Anti-ANTXR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306122EA 600 EUR
ORIGTA306122
Anti-ANTXR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ANTXR1 (anthrax toxin receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR