Anti-TIRAP Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306132EA 600 EUR
ORIGTA306132
Anti-TIRAP Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 4 ?g/ml

Type: Primary
Antigen: TIRAP (toll-interleukin 1 receptor (TIR) domain containing adaptor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR