Anti-BCL2L14 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306135EA 595 EUR
ORIGTA306135
Anti-BCL2L14 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2.5-5 ?g/ml

Type: Primary
Antigen: BCL2L14 (BCL2-like 14 (apoptosis facilitator))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR