Anti-CUL9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306136EA 600 EUR
ORIGTA306136
Anti-CUL9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 1 ?g/ml; Immunofluorescence: 2 ?g/ml

Type: Primary
Antigen: CUL9 (cullin 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR