Drucken

Anti-CARD11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306139EA 482 EUR InStock
ORIGTA306139
Anti-CARD11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: CARD11 (caspase recruitment domain family, member 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more