Anti-BAD Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306174EA 600 EUR
ORIGTA306174
Anti-BAD Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR